Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RabX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132399.3 | NP_572627.1 | 1 | 607 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |