Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042804.3 | NP_001036269.2 | 5 - 6 | 8515 | 103 | |
NM_001272506.1 | NP_001259435.1 | 2 - 3 | 3124 | 103 | ||
NM_001298174.1 | NP_001285103.1 | 4 - 5 | 8339 | 103 | ||
NM_132441.3 | NP_572669.2 | 4 - 5 | 8242 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |