Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: antdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132467.3 | NP_572695.2 | 1 - 2 | 604 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |