Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp28c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272535.1 | NP_001259464.1 | 1 - 2 | 187 | 73 | |
NM_132524.2 | NP_572752.1 | 1 - 2 | 187 | 73 | ||
GenBank mRNA | BT025008.1 | - | 1 - 2 | 187 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |