Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp28c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272535.1 | NP_001259464.1 | 3 - 4 | 944 | 66 | |
NM_132524.2 | NP_572752.1 | 3 - 4 | 944 | 66 | ||
GenBank mRNA | BT025008.1 | - | 3 - 4 | 944 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |