Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wisp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298214.1 | NP_001285143.1 | 4 - 5 | 3093 | 65 | |
NM_001298215.1 | NP_001285144.1 | 3 - 4 | 3289 | 65 | ||
NM_132538.2 | NP_572766.1 | 4 - 5 | 3096 | 65 | ||
GenBank mRNA | BT149893.1 | - | 3 - 4 | 3287 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |