Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103493.4 | NP_001096963.1 | 10 - 11 | 6254 | 80 | |
NM_001298228.1 | NP_001285157.1 | 11 - 12 | 6721 | 80 | ||
NM_001298230.1 | NP_001285159.1 | 12 - 13 | 7036 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |