Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tango2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258725.2 | NP_001245654.1 | 4 - 5 | 942 | 75 | |
NM_001272596.1 | NP_001259525.1 | 4 - 5 | 693 | 75 | ||
NM_001298288.1 | NP_001285217.1 | 3 - 4 | 1013 | 75 | ||
NM_132677.4 | NP_572905.1 | 3 - 4 | 842 | 75 | ||
GenBank mRNA | AY069312.1 | - | 3 - 4 | 814 | 75 | |
BT089002.1 | - | 3 - 4 | 977 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |