Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201657.2 | NP_001188586.1 | 2 - 3 | 1813 | 71 | |
NM_001201658.2 | NP_001188587.1 | 2 - 3 | 1813 | 71 | ||
NM_001298303.1 | NP_001285232.1 | 2 - 3 | 1813 | 71 | ||
NM_132702.5 | NP_572930.3 | 3 - 4 | 1843 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |