Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsd-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042811.2 | NP_001036276.1 | 1 - 2 | 472 | 63 | |
NM_001042812.2 | NP_001036277.2 | 1 - 2 | 472 | 63 | ||
NM_001298332.1 | NP_001285261.1 | 1 - 2 | 472 | 63 | ||
NM_001298333.1 | NP_001285262.1 | 1 - 2 | 472 | 63 | ||
NM_132768.3 | NP_572996.1 | 1 - 2 | 472 | 63 | ||
GenBank mRNA | AY060242.1 | - | 1 - 2 | 455 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |