Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGRP-LE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258766.3 | NP_001245695.1 | 4 - 5 | 1551 | 67 | |
NM_132850.4 | NP_573078.1 | 3 - 4 | 876 | 67 | ||
GenBank mRNA | AF313391.1 | - | 3 - 4 | 833 | 67 | |
AY058258.1 | - | 3 - 4 | 832 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |