Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CalpC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132890.3 | NP_573118.2 | 1 - 2 | 909 | 65 | |
GenBank mRNA | BT150312.1 | - | 1 - 2 | 861 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |