Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272701.1 | NP_001259630.1 | 2 - 3 | 167 | 84 | |
NM_132948.4 | NP_573176.1 | 1 - 2 | 207 | 84 | ||
GenBank mRNA | AY060757.1 | - | 1 - 2 | 186 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |