Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: X11L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132996.4 | NP_573224.2 | 3 - 4 | 1143 | 86 | |
GenBank mRNA | AY051760.1 | - | 3 - 4 | 1110 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |