Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: X11L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169310.2 | NP_001162781.1 | 9 - 10 | 3498 | 65 | |
NM_001272725.2 | NP_001259654.1 | 9 - 10 | 3857 | 65 | ||
NM_001272726.2 | NP_001259655.2 | 9 - 10 | 3480 | 65 | ||
NM_001298447.1 | NP_001285376.1 | 10 - 11 | 3504 | 65 | ||
NM_132996.4 | NP_573224.2 | 9 - 10 | 3495 | 65 | ||
GenBank mRNA | AY051760.1 | - | 9 - 10 | 3462 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |