Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014557.2 | NP_001014557.3 | 3 - 4 | 1752 | 88 | |
NM_001169873.1 | NP_001163344.1 | 3 - 4 | 1752 | 88 | ||
NM_001274478.1 | NP_001261407.1 | 3 - 4 | 1752 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |