Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Teh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139609.2 | NP_647866.2 | 6 - 7 | 1736 | 62 | |
GenBank mRNA | BT022560.1 | - | 6 - 7 | 1133 | 62 | |
DQ139960.1 | - | 6 - 7 | 1109 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |