Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Teh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169874.3 | NP_001163345.1 | 4 - 5 | 2069 | 62 | |
NM_139610.4 | NP_647867.1 | 3 - 4 | 681 | 62 | ||
GenBank mRNA | BT050528.1 | - | 4 - 5 | 2070 | 62 | |
DQ139958.1 | - | 3 - 4 | 646 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |