Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DopEcR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014559.3 | NP_001014559.1 | 2 - 3 | 1491 | 65 | |
NM_001014560.2 | NP_001014560.1 | 3 - 4 | 1945 | 65 | ||
NM_139640.3 | NP_647897.2 | 3 - 4 | 1567 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Drosophila melanogaster |