Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LeuRS-m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139675.3 | NP_647932.1 | 1 - 2 | 186 | 62 | |
GenBank mRNA | AY089557.1 | - | 1 - 2 | 142 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |