Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rh50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274505.2 | NP_001261434.1 | 1 - 2 | 492 | 90 | |
NM_001300009.1 | NP_001286938.1 | 1 - 2 | 492 | 90 | ||
NM_001382192.1 | NP_001369039.1 | 1 - 2 | 492 | 90 | ||
NM_139683.3 | NP_647940.1 | 1 - 2 | 492 | 90 | ||
GenBank mRNA | AF172639.1 | - | 1 - 2 | 474 | 90 | |
AF181624.1 | - | 1 - 2 | 474 | 90 | ||
AF193812.1 | - | 1 - 2 | 474 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |