Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gen | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139686.3 | NP_647943.2 | 2 - 3 | 537 | 66 | |
GenBank mRNA | AY128475.1 | - | 2 - 3 | 508 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |