Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eaf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139724.4 | NP_647981.1 | 3 - 4 | 557 | 76 | |
GenBank mRNA | AY071012.1 | - | 3 - 4 | 498 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |