Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jon65Aiv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139755.3 | NP_648012.1 | 1 - 2 | 299 | 59 | |
GenBank mRNA | AY061559.1 | - | 1 - 2 | 509 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |