Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zpg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274549.1 | NP_001261478.1 | 1 - 2 | 398 | 99 | |
NM_139792.3 | NP_648049.1 | 1 - 2 | 398 | 99 | ||
GenBank mRNA | AF271718.1 | - | 1 - 2 | 364 | 99 | |
AY094856.1 | - | 1 - 2 | 395 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |