Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139833.2 | NP_648090.1 | 2 - 3 | 1358 | 62 | |
GenBank mRNA | AY119656.1 | - | 2 - 3 | 1305 | 62 | |
BT088971.1 | - | 3 - 4 | 2278 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |