Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sarm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043129.3 | NP_001036594.1 | 3 - 4 | 1418 | 57 | |
NM_001104071.2 | NP_001097541.1 | 4 - 5 | 1566 | 57 | ||
NM_001104074.3 | NP_001097544.1 | 5 - 6 | 1500 | 57 | ||
NM_001104075.2 | NP_001097545.1 | 4 - 5 | 1324 | 57 | ||
NM_001259733.1 | NP_001246662.1 | 6 - 7 | 3111 | 57 | ||
NM_001259734.1 | NP_001246663.1 | 3 - 4 | 1118 | 57 | ||
NM_001274627.1 | NP_001261556.1 | 5 - 6 | 2524 | 57 | ||
GenBank mRNA | BT014666.1 | - | 4 - 5 | 1298 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |