Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exo70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139965.3 | NP_648222.3 | 3 - 4 | 835 | 94 | |
GenBank mRNA | BT133119.1 | - | 3 - 4 | 765 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Drosophila melanogaster |