Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144440.2 | NP_001137912.1 | 5 - 6 | 5338 | 85 | |
NM_001259740.2 | NP_001246669.1 | 7 - 8 | 5383 | 85 | ||
NM_001274650.1 | NP_001261579.1 | 5 - 6 | 2321 | 85 | ||
NM_001300054.1 | NP_001286983.1 | 5 - 6 | 5338 | 85 | ||
NM_139969.4 | NP_648226.2 | 7 - 8 | 5344 | 85 | ||
GenBank mRNA | BT133161.1 | - | 5 - 6 | 2317 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |