Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144440.2 | NP_001137912.1 | 8 - 9 | 6631 | 66 | |
NM_001259740.2 | NP_001246669.1 | 10 - 11 | 6676 | 66 | ||
NM_001274650.1 | NP_001261579.1 | 8 - 9 | 3614 | 66 | ||
NM_001300054.1 | NP_001286983.1 | 8 - 9 | 6631 | 66 | ||
NM_139969.4 | NP_648226.2 | 10 - 11 | 6637 | 66 | ||
GenBank mRNA | BT133161.1 | - | 8 - 9 | 3610 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |