Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259745.2 | NP_001246674.1 | 14 - 15 | 7473 | 101 | |
NM_001274657.1 | NP_001261586.1 | 13 - 14 | 7826 | 101 | ||
NM_001274658.1 | NP_001261587.1 | 14 - 15 | 7410 | 101 | ||
NM_001274659.1 | NP_001261588.1 | 14 - 15 | 7595 | 101 | ||
NM_001274660.1 | NP_001261589.1 | 15 - 16 | 7611 | 101 | ||
NM_139981.4 | NP_648238.1 | 13 - 14 | 7298 | 101 | ||
GenBank mRNA | AF299248.1 | - | 12 - 13 | 7245 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |