Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259745.2 | NP_001246674.1 | 3 - 4 | 589 | 90 | |
NM_001259746.2 | NP_001246675.1 | 3 - 4 | 589 | 90 | ||
NM_001259747.2 | NP_001246676.1 | 3 - 4 | 701 | 90 | ||
NM_001274657.1 | NP_001261586.1 | 2 - 3 | 942 | 90 | ||
NM_001274658.1 | NP_001261587.1 | 3 - 4 | 589 | 90 | ||
NM_001274659.1 | NP_001261588.1 | 3 - 4 | 711 | 90 | ||
NM_001274660.1 | NP_001261589.1 | 3 - 4 | 701 | 90 | ||
NM_139981.4 | NP_648238.1 | 2 - 3 | 414 | 90 | ||
GenBank mRNA | AF299248.1 | - | 1 - 2 | 361 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |