Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273182.1 | NP_001260111.1 | 4 - 5 | 1271 | 70 | |
NM_001273183.2 | NP_001260112.1 | 4 - 5 | 1271 | 70 | ||
NM_001273184.2 | NP_001260113.1 | 4 - 5 | 1271 | 70 | ||
NM_057217.4 | NP_476565.1 | 4 - 5 | 949 | 70 | ||
NM_057218.4 | NP_476566.1 | 4 - 5 | 949 | 70 | ||
NM_057219.4 | NP_476567.1 | 4 - 5 | 949 | 70 | ||
GenBank mRNA | BT044153.1 | - | 4 - 5 | 935 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |