Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: her | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273577.1 | NP_001260506.1 | 2 - 3 | 1327 | 63 | |
NM_057224.5 | NP_476572.1 | 2 - 3 | 1327 | 63 | ||
GenBank mRNA | AY058630.1 | - | 2 - 3 | 1292 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |