Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043090.3 | NP_001036555.1 | 3 - 4 | 561 | 73 | |
NM_001043091.3 | NP_001036556.1 | 3 - 4 | 574 | 73 | ||
NM_001043092.3 | NP_001036557.1 | 3 - 4 | 561 | 73 | ||
NM_057227.5 | NP_476575.1 | 3 - 4 | 561 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |