Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057251.4 | NP_476599.1 | 1 - 2 | 750 | 84 | |
GenBank mRNA | AF145679.1 | - | 1 - 2 | 651 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |