Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lace | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299028.1 | NP_001285957.1 | 3 - 4 | 1674 | 61 | |
NM_057266.5 | NP_476614.1 | 3 - 4 | 1674 | 61 | ||
GenBank mRNA | AY095191.1 | - | 3 - 4 | 1644 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |