Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mal-A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057277.3 | NP_476625.2 | 2 - 3 | 1648 | 81 | |
GenBank mRNA | AY071566.1 | - | 2 - 3 | 1642 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |