Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Appl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258519.1 | NP_001245448.1 | 6 - 7 | 1447 | 68 | |
NM_001258520.2 | NP_001245449.1 | 5 - 6 | 1290 | 68 | ||
NM_057278.5 | NP_476626.2 | 6 - 7 | 1461 | 68 | ||
GenBank mRNA | AF181628.1 | - | 5 - 6 | 1436 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |