Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274138.2 | NP_001261067.1 | 2 - 3 | 2151 | 70 | |
NM_057324.6 | NP_476672.1 | 2 - 3 | 2151 | 70 | ||
GenBank mRNA | AY075585.1 | - | 2 - 3 | 2123 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |