Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057354.3 | NP_476702.1 | 8 - 9 | 1748 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |