Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RanGAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273649.1 | NP_001260578.1 | 4 - 5 | 1600 | 64 | |
NM_001273650.1 | NP_001260579.1 | 5 - 6 | 1736 | 64 | ||
NM_057364.4 | NP_476712.1 | 5 - 6 | 1993 | 64 | ||
GenBank mRNA | AY061219.1 | - | 4 - 5 | 1585 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |