Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tud | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299736.1 | NP_001286665.1 | 6 - 7 | 4102 | 65 | |
NM_057425.3 | NP_476773.1 | 6 - 7 | 4111 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |