Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tup | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299139.1 | NP_001286068.1 | 7 - 8 | 1615 | 56 | |
NM_057426.4 | NP_476774.1 | 7 - 8 | 1615 | 56 | ||
NM_057427.3 | NP_476775.1 | 7 - 8 | 1615 | 56 | ||
GenBank mRNA | AF145674.1 | - | 7 - 8 | 1600 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |