Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbp53E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299582.1 | NP_001286511.1 | 4 - 5 | 553 | 80 | |
NM_057490.4 | NP_476838.1 | 3 - 4 | 504 | 80 | ||
GenBank mRNA | AY058340.1 | - | 3 - 4 | 471 | 80 | |
X68566.1 | - | 453 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |