Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: retn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299844.1 | NP_001286773.1 | 9 - 10 | 2302 | 58 | |
NM_001299845.1 | NP_001286774.1 | 5 - 6 | 1068 | 58 | ||
NM_001299846.1 | NP_001286775.1 | 5 - 6 | 1552 | 58 | ||
NM_057516.4 | NP_476864.2 | 9 - 10 | 2308 | 58 | ||
NM_176254.2 | NP_788434.1 | 9 - 10 | 2323 | 58 | ||
GenBank mRNA | AY118955.1 | - | 9 - 10 | 2292 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |