Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: piwi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298896.1 | NP_001285825.1 | 1 - 2 | 176 | 95 | |
NM_057527.4 | NP_476875.1 | 1 - 2 | 176 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |