Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: noc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299003.1 | NP_001285932.1 | 1 - 2 | 480 | 87 | |
NM_057569.3 | NP_476917.1 | 1 - 2 | 480 | 87 | ||
GenBank mRNA | AY070550.1 | - | 1 - 2 | 450 | 87 | |
L14009.1 | - | 1 - 2 | 477 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |