Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sgl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057632.4 | NP_476980.1 | 2 - 3 | 497 | 72 | |
GenBank mRNA | AF000570.1 | - | 2 - 3 | 499 | 72 | |
AF009013.1 | - | 2 - 3 | 493 | 72 | ||
AY052137.1 | - | 2 - 3 | 480 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |