Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: babo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169621.2 | NP_001163092.1 | 6 - 7 | 1717 | 76 | |
NM_057651.4 | NP_476999.1 | 6 - 7 | 1798 | 76 | ||
NM_057652.4 | NP_477000.1 | 6 - 7 | 1735 | 76 | ||
GenBank mRNA | BT009976.1 | - | 6 - 7 | 1715 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |